missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NET1 Antibody (CL3063) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46648JF549
This item is not returnable.
View return policy
Description
NET1 Monoclonal antibody specifically detects NET1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| NET1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| ARHGEF8neuroepithelial cell-transforming gene 1 protein, guanine nucleotide regulatory protein (oncogene), NET1A, neuroepithelial cell transforming 1, neuroepithelial cell transforming gene 1, neuroepithelioma transforming gene 1, p65 Net1 proto-oncogene protein, Proto-oncogene p65 Net1, Rho guanine nucleotide exchange factor (GEF) 8, Rho guanine nucleotide exchange factor 8, small GTP-binding protein regulator | |
| This antibody was developed using a recombinant protein derived from P23975, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3063 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 10276 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction