missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurobeachin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 589.00 €
Specifications
| Antigen | Neurobeachin |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18463642
|
Novus Biologicals
NBP1-90003-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18295666
|
Novus Biologicals
NBP1-90003 |
0.1 mL |
589.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurobeachin Polyclonal specifically detects Neurobeachin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Neurobeachin | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 26960 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FKIHTTSDGMSSISERDLASSTKGLEYAEMTATTLETESSSSKIVPNIDAGSIISDTERSDDGKESGKEIRKIQTTTTTQAVQGRSITQQDRDLRVDLGFR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BCL8Blysosomal trafficking regulator 2, EC 1.14.14.5, EC 6.1.1.11, FLJ10197, KIAA1544, Lysosomal-trafficking regulator 2, LYST2, neurobeachin, Protein BCL8B | |
| NBEA | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title