missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeuroD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585775
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%). Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse fetal brain, rat cerebellum. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NeuroD1 is a transcriptional activator that acts as a differentiation factor during neurogenesis. It has been demonstrated to bind to the insulin gene E-box. Efficient DNA binding requires dimerization with another basic helix-loop-helix (bHLH) protein. Defects in NEUROD1 are a cause of maturity onset diabetes of the young type VI (MODY6). MODY6 is a form of non-insulin-dependent diabetes mellitus characterized by an autosomal dominant mode of inheritance, onset during young adulthood and a primary defect in insulin secretion.
Specifications
| NeuroD1 | |
| Polyclonal | |
| Unconjugated | |
| NEUROD1 | |
| Basic helix-loop-helix factor 1; basic helix-loop-helix transcription factor; Beta2; beta-cell E-box transactivator 2; beta-cell E-box transcriptional activator 2; BHF1; BHF-1; bHLHa3; Class A basic helix-loop-helix protein 3; MODY6; neuro d1; Neurod; Neurod1; neurogenic differentiation 1; neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD; neuronal differentiation 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18012, 29458, 4760 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.63 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q13562, Q60867, Q64289 | |
| NEUROD1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse NeuroD1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction