missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeuroD2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148627
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Members of the myogenic determination family are basic helix-loop-helix (bHLH) proteins that can be separated into two classes, both of which work together to activate DNA transcription. Class A proteins include the ubiquitously expressed E-box binding factors, namely E2A, ITF-2 and HEB, while class B proteins, such as MyoD, myogenin and Neuro D (BETA2), are transiently expressed and exhibit a much more limited tissue distribution. Neuro D2 (neurogenic differentiation 2), also known as NDRF, NEUROD2 or bHLHa1, is a 382 amino acid nuclear protein that contains one bHLH domain and functions to induce neurogenic differentiation, playing an important role in the maintenance and determination of cell fate.
Specifications
| NeuroD2 | |
| Polyclonal | |
| Unconjugated | |
| NEUROD2 | |
| BHLHA1; Brain bHLH protein KW8; class A basic helix-loop-helix protein 1; NDF2; NDR2; NDRF; NeuroD2; neuroD-related factor; neurogenic basic-helix-loop-helix protein; neurogenic differentiation 2; neurogenic differentiation factor 2; neuronal differentiation 2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 18013, 4761 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q15784, Q62414 | |
| NEUROD2 | |
| E. coli-derived recombinant human NeuroD2, Ser174-Asn382. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction