Learn More
Neurofibromin 1 Antibody (CL11865) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15123FR
Product Code. 30290890
  • 737.00 € / 0.10mL
Description
Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neurofibromin 1
Monoclonal
DyLight 680
DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
This antibody was developed using a synthetic peptide derived from P21359, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
4763
Store at 4⪚C in the dark.
IgG2b
Immunocytochemistry/Immunofluorescence
CL11865
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified