missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00 € - 372.00 €
Specifications
| Antigen | Neurogenin-3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18199053
|
Novus Biologicals
NBP2-27115SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18797983
|
Novus Biologicals
NBP2-27115 |
0.1 mg |
372.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin-3 Polyclonal specifically detects Neurogenin-3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Neurogenin-3 | |
| Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Rabbit | |
| Stem Cells | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 50674 | |
| A portion of amino acids 1-69 of mouse Neurog 3 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
| NEUROG3 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 83% homologous in rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title