missing translation for 'onlineSavingsMsg'
Learn More
Learn More
neuron navigator 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
NAV1 belongs to the neuron navigator family and is expressed predomitly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. The exact function of this gene is not known, but it is thought to play a role in in neuronal development and regeneration. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | neuron navigator 1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NAV1 |
| Gene Accession No. | Q8CH77, Q8NEY1 |
| Gene Alias | KIAA1151, KIAA1213, NAV1, neuron navigator 1, POMFIL3, steerin 1, STEERIN1, Unc 53 homolog 1, unc53H1 |
| Gene Symbols | NAV1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?