missing translation for 'onlineSavingsMsg'
Learn More

neuron navigator 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16810395 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16810395 20 μL 20µL
16800395 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16810395 Supplier Proteintech Supplier No. 208071AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAV1 belongs to the neuron navigator family and is expressed predomitly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. The exact function of this gene is not known, but it is thought to play a role in in neuronal development and regeneration. Alternative splicing results in multiple transcript variants.

Specifications

Antigen neuron navigator 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NAV1
Gene Accession No. Q8CH77, Q8NEY1
Gene Alias KIAA1151, KIAA1213, NAV1, neuron navigator 1, POMFIL3, steerin 1, STEERIN1, Unc 53 homolog 1, unc53H1
Gene Symbols NAV1
Host Species Rabbit
Immunogen neuron navigator 1 Fusion Protein Ag14747
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215690, 685707, 89796
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.