missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuron-specific beta-III Tubulin NL637 Antibody (Clone TuJ-1), R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems NL1195V
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Multi-Species samples. It is validated for Immunocytochemistry.
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| NorthernLights 637 (NL637) | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| TUBB3 | |
| Rat brain-derived microtubules. | |
| 0.5 mL | |
| Primary | |
| Detects neuron-specific beta-III Tubulin in Western blots. | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. | |
| IgG2a |
| Immunocytochemistry | |
| TuJ-1 | |
| Immunocytochemistry | |
| Beta3-tubulin, beta-4, betaIII Tubulin, beta-Tubulin III, Class III beta-tubulin, TUBB3, TUBB4 CFEOM3A, Tubulin Beta 3, tubulin beta-3 chain, Tubulin beta-4, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10381 | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction