missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuroserpin Monoclonal Antibody (437016)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561263
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. The protein is primarily secreted by axons in the brain, and preferentially reacts with and inhibits tissue-type plasminogen activator. It is thought to play a role in the regulation of axonal growth and the development of synaptic plasticity. Mutations in this gene result in familial encephalopathy with neuroserpin inclusion bodies (FENIB), which is a dominantly inherited form of familial encephalopathy and epilepsy characterized by the accumulation of mutant neuroserpin polymers. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| Neuroserpin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O35684, Q99574 | |
| Serpini1 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin I1, Ala19-Leu410. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 437016 | |
| Unconjugated | |
| Serpini1 | |
| AI837402; DKFZp781N13156; neuroserpin; Ns; Peptidase inhibitor 12; PI12; PI-12; protease inhibitor 17; raPIT5a; serine (or cysteine) peptidase inhibitor, clade I, member 1; serine (or cysteine) proteinase inhibitor clade member 1; serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1; serine protease inhibitor 17; serpin family I member 1; serpin I1; serpin peptidase inhibitor clade I member 1; serpin peptidase inhibitor, clade I (neuroserpin), member 1; serpin peptidase inhibitor, clade I, member 1; SERPINI1; Spi17 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20713, 5274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction