missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophils Antibody (RPN3/57), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11629-0.025mg
This item is not returnable.
View return policy
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Neutrophils | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Rabbit | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| RPN3/57 | |
| PBS | |
| Rabbit Peritoneal Neutrophils | |
| 0.025 mg | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction