missing translation for 'onlineSavingsMsg'
Learn More

Nevada Pesticide Mixture Version 2 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321986
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321986 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321986 Supplier Dr. Ehrenstorfer missing translation for 'supplierNo' DREGS09000694AL
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abamectin∣Acequinocyl∣Bifenazate∣Bifenthrin∣Cyfluthrin∣Cypermethrin∣Daminozide∣Dimethomorph∣Etoxazole∣Fenhexamid∣Flonicamid∣Fludioxonil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Piperonyl butoxide∣Pyrethrins∣Quintozene∣Spinetoram J∣Spinosad∣Spirotetramat∣Thiamethoxam∣Trifloxystrobin
CAS 71751-41-2, 57960-19-7, 149877-41-8, 82657-04-3, 68359-37-5, 52315-07-8
Analyte or Component Names Abamectin∣Acequinocyl∣Bifenazate∣Bifenthrin∣Cyfluthrin∣Cypermethrin∣Daminozide∣Dimethomorph∣Etoxazole∣Fenhexamid∣Flonicamid∣Fludioxonil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Piperonyl butoxide∣Pyrethrins∣Quintozene∣Spinetoram J∣Spinosad∣Spirotetramat∣Thiamethoxam∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Abamectin 100 μg/mL∣Acequinocyl 100 μg/mL∣Bifenazate 100 μg/mL∣Bifenthrin 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Daminozide 100 μg/mL∣Dimethomorph 100 μg/mL∣Etoxazole 100 μg/mL∣Fenhexamid 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL∣Imidacloprid 100 μg/mL∣Myclobutanil 100 μg/mL∣Paclobutrazol 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Pyrethrins 100 μg/mL∣Quintozene 100 μg/mL∣Spinetoram J 100 μg/mL∣Spinosad 100 μg/mL∣Spirotetramat 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.