missing translation for 'onlineSavingsMsg'
Learn More

Nevada Terpene Mixture 0058 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer

Product Code. 30316894
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316894 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316894 Supplier Dr. Ehrenstorfer Supplier No. DREA50000058IP
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Trans-Caryophyllene∣alpha-Bisabolol∣alpha-Humulene∣beta-Pinene∣Caryophyllene Oxide∣DL-alpha-Pinene∣DL-Linalool∣Limonene∣Myrcene∣Terpinolene
CAS 87-44-5, 515-69-5, 6753-98-6, 127-91-3, 1139-30-6, 80-56-8
Analyte or Component Names (-)-Trans-Caryophyllene∣alpha-Bisabolol∣alpha-Humulene∣beta-Pinene∣Caryophyllene Oxide∣DL-alpha-Pinene∣DL-Linalool∣Limonene∣Myrcene∣Terpinolene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Isopropanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (-)-Trans-Caryophyllene 1000 mg/L∣alpha-Bisabolol 1000 mg/L∣alpha-Humulene 1000 mg/L∣beta-Pinene 1000 mg/L∣Caryophyllene Oxide 1000 mg/L∣DL-alpha-Pinene 1000 mg/L∣DL-Linalool 1000 mg/L∣Limonene 1000 mg/L∣Myrcene 1000 mg/L∣Terpinolene 1000 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.