missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Newcastle Disease Virus (NDV) Antibody (9F7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-59912AF594
This item is not returnable.
View return policy
Description
Newcastle Disease Virus (NDV) Monoclonal antibody specifically detects Newcastle Disease Virus (NDV) in Virus samples. It is validated for Western Blot, Haemagglutination, ELISA
Specifications
| Newcastle Disease Virus (NDV) | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Hemagglutination Assay, ELISA | |
| 9F7 | |
| Western Blot, Haemagglutination, Sandwich ELISA | |
| NDV | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Newcastle disease virus (La-sota strain). | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction