Learn More
Nexilin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27737-1-AP-20UL
 View more versions of this product
Product Code. 18026705
  • 145.00 € / 20µL
Description
The Nexilin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Nexilin. This antibody recognizes human, mouse, rat antigen. The Nexilin antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Nexilin
650 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
F actin binding protein, NELIN, Nexilin, NEXN
20 μL
91624
Store at -20°C. Stable for one year after shipment.