missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NF-M Recombinant Mouse Monoclonal Antibody (K1E065_3G3)
GREENER_CHOICE

Product Code. 30824789
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824789 100 μL
30830372 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824789 Supplier Invitrogen™ Supplier No. MA564444

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: EKEQLQGLND RFAGYIEKVH YLEQQNKEIE AEIQALRQKQ ASHAQLGDAY DQEIRELRAT LEMVNHEKAQ VQLDSDHLEE DIHRLKERFE EEARLRDDTE AAIRALRKDI EEASLVKVEL DKKVQSLQDE VAFLRSNHEE EVADLLAQIQ ASHITVERKD YLKTDISTAL KEIRSQLESH SDQNMHQAEE WFKCRYAKLT EAAEQNKEAI RSAKEEIAEY RRQLQSKSIE LESVRGTKES LERQLSDIEE RHNHDLSSYQ DTIQQLENEL RGTKWEMARH LREYQDLLNV KMALDIEIAA YRKLLEGEET RFSTFAGSIT GPLYTHRPPI TISSKIQKPK VEAPKLKVQH KFVEEIIEET KVEDEKSEME EALTAITEEL AVSMKEEKKE AAEEKEEEPE AEEEEVAAKK SPVKATAPEV KEEEGEKEEE EGQEEEEEED EGAKSDQAEE GGSEKEGSSE KEEGEQEEGE TEAEAEGEEA EAKEEKKVEE KSEEVATKEE LVADAKVEKP EKAKSPVPKS PVEEKGKSPV PKSPVEEKGK SPVPKSPVEE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%, Pig - 90%, Cynomolgus monkey - 96%.

Neurofilaments (NFs) are a type of intermediate filament (IF) expressed almost exclusively in neuronal cells, and in those cells most prominently in large axons. NFs, in most vertebrates, are composed of three different polypeptide chains with different molecular weights - neurofilament medium protein (NF-M), high (NF-H) and light protein (NF-L), which share sequence and structural similarity in a coiled-coil core domain, but differ in the length and sequence of their N-termini and more dramatically of their C-termini which in the case of NF-M and NF-H form the flexible extensions that link NFs to each other and to other elements in the cytoplasm. NF-M protein tail-mediated interactions of neurofilaments are critical for size and cytoskeletal architecture of axons, and are mediated, in part, by the highly phosphorylated tail domain of this protein. NF-M phosphorylation and O-GlcNAcylation are regulated reciprocally and affect its translocation and filament formation and function. Antibodies to the various neurofilament subunits are very useful cell type markers since the proteins are among the most abundant of the nervous system, are expressed only in neurons and are biochemically very stable.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-M
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K1E065_3G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene NEFM
Gene Accession No. P07197
Gene Alias 160 kDa neurofilament protein; major neuronal intermediate filament subunit; medium neurofilament subunit; middle molecular weight neurofilament protein NF-M(2); Nef3; Nefm; nef-m; nefm.S; nefm-a; nefm-b; Neurofilament 3; neurofilament 3 (150kDa medium); neurofilament 3, medium; neurofilament M subunit; neurofilament medium; neurofilament medium polypeptide; neurofilament medium S homeolog; neurofilament medium tail domain; neurofilament protein M; Neurofilament protein, middle polypeptide; neurofilament triplet M protein; neurofilament, medium polypeptide; neurofilament, medium polypeptide 150kDa; neurofilament, medium polypeptide S homeolog; neurofilament-3 (150 kD medium); neurofilament-M; neurofilament-M subunit; neuronal intermediate filament; NF160; NF165; NFM; NF-M; NF-M c-terminus; NF-M protein; NMC; XELAEV_18020229mg
Gene Symbols NEFM
Host Species Mouse
Immunogen Recombinant human NEFM (aa 101-650).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4741
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.