missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NF1 Monoclonal Antibody (McNFn27a)
GREENER_CHOICE

Product Code. 11508852
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11508852 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11508852 Supplier Invitrogen™ Supplier No. MA1084

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA1-084 detects neurofibromin in human, rat and mouse cells. MA1-084 has been successfully used in Western blot, procedures. By Western blot, this antibody detects a ~230-260 kDa protein. The MA1-084 immunogen is a synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin.

The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen NF1
Applications Affinity Purification, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone McNFn27a
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene NF1
Gene Accession No. P21359, P97526, Q04690
Gene Alias AW494271; DKFZp686J1293; E030030H24Rik; FLJ21220; neurofibromatosis 1; Neurofibromatosis type 1; neurofibromatosis-related protein NF-1; Neurofibromin; neurofibromin 1; Neurofibromin truncated; NF1; Nf-1; NFNS; VRNF; WSS
Gene Symbols NF1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18015, 24592, 4763
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.