missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF2/Merlin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
353.00 €
Specifications
| Antigen | NF2/Merlin |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435821
|
Novus Biologicals
NBP1-87757-25ul |
25 μL |
353.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039043
|
Novus Biologicals
NBP1-87757 |
0.1 mL |
N/A
|
N/A | |||||
Description
NF2/Merlin Polyclonal specifically detects NF2/Merlin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| NF2/Merlin | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4771 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ERRAKQKLLEIATKPTYPPMNPIPAPLPPDIPSFNLIGDSLSFDFKDTDMKRLSMEIEKEKVEYMEKSKHLQEQLNELKTEIEALKLKERETALDILHNENSDRGGSSKHNTIKKLTLQSAKSRVA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ACN, BANF, merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
| NF2 | |
| IgG | |
| Affinity Purified | |
| Specificity of human NF2/Merlin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title