missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFAM1 Monoclonal Antibody (374301)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561376
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells. It is also known CNAIP.
Specifications
| NFAM1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8NET5 | |
| NFAM1 | |
| E. coli-derived recombinant human NFAM1, Gln43-Lys163. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 374301 | |
| Unconjugated | |
| NFAM1 | |
| 4921501M20Rik; bK126B4.4; Calcineurin/NFAT-activating ITAM-containing protein; Cnaip; CTA-126B4.4; Nfam1; Nfat activating molecule with ITAM motif 1; NFAT activating protein with ITAM motif 1; NFAT activation molecule 1; NFAT-activating protein with ITAM motif 1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 150372 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction