missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC2/NFAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
396.00 € - 513.00 €
Specifications
| Antigen | NFATC2/NFAT1 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18492911
|
Novus Biologicals
NBP1-82583-25ul |
25 μL |
396.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18453371
|
Novus Biologicals
NBP1-82583 |
0.1 mL |
513.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFATC2/NFAT1 Polyclonal antibody specifically detects NFATC2/NFAT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NFATC2/NFAT1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Transcription Factors and Regulators, Wnt Signaling Pathway | |
| PBS (pH 7.2) and 40% Glycerol | |
| 4773 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DFSILFDYEYLNPNEEEPNAHKVASPPSGPAYPDDVLDYGLKPYSPLASLSGEPPGRFGEPDRVGPQKFLSAAKPAGASGLSPRIEITPSHELIQAVGPLRMRDAGLLVEQPPLAGVAASPRFTLPV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title