missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB2/NFkB p100 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
235.00 € - 483.00 €
Specifications
| Antigen | NFkB2/NFkB p100 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18300087
|
Novus Biologicals
NBP3-03791-20UL |
20 μL |
235.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18361016
|
Novus Biologicals
NBP3-03791-100UL |
100 μL |
483.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFkB2/NFkB p100 Polyclonal specifically detects NFkB2/NFkB p100 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| NFkB2/NFkB p100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 690-899 of human NFkB2/NFkB p100 (NP_002493.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 4791 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title