missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93532-0.1ml
This item is not returnable.
View return policy
Description
NFKBIL1 Polyclonal antibody specifically detects NFKBIL1 in Human samples. It is validated for Western Blot
Specifications
| NFKBIL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| IkappaBL, I-kappa-B-like protein, IKBLikappaBL, Inhibitor of kappa B-like protein, LST1, NF-kappa-B inhibitor-like protein 1, nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human NFKBIL1 (NP_004998.3). MSNPSPQVPEEEASTSVCRPKSSMASTSRRQRRERRFRRYLSAGRLVRAQALLQRHPGLDVDAGQPPPLHRACARHDAPALCLLLRLGADPAHQDRHGDTALHAAARQGPDAYTDFFLPLLSRCPSAMGIKNKDGETPGQILGWGPPWDSAEEEEEDDASKEREWRQKLQGELEDEWQEVMGRFEGDASHETQEPESFSAWSDRLAREHAQKCQQQQREAEGSRRPPRAEGSSQSWRQQEEEQRLFRERARAKEEELRESRARRAQEALGDREPKPTRAGPREEHPRGAGRGSLWRFGDVPWPCPGGGDPEAMAAALVARGPPLEEQGALRRYLRVQQVRWHPDRFLQRFRSQIETWELGRVMGAVTALSQALNRHAEALK | |
| 0.1 mL | |
| Signal Transduction | |
| 4795 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction