missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | NFKBIL1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NFKBIL1 Polyclonal antibody specifically detects NFKBIL1 in Human samples. It is validated for Western BlotSpecifications
| NFKBIL1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4795 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| IkappaBL, I-kappa-B-like protein, IKBLikappaBL, Inhibitor of kappa B-like protein, LST1, NF-kappa-B inhibitor-like protein 1, nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human NFKBIL1 (NP_004998.3). MSNPSPQVPEEEASTSVCRPKSSMASTSRRQRRERRFRRYLSAGRLVRAQALLQRHPGLDVDAGQPPPLHRACARHDAPALCLLLRLGADPAHQDRHGDTALHAAARQGPDAYTDFFLPLLSRCPSAMGIKNKDGETPGQILGWGPPWDSAEEEEEDDASKEREWRQKLQGELEDEWQEVMGRFEGDASHETQEPESFSAWSDRLAREHAQKCQQQQREAEGSRRPPRAEGSSQSWRQQEEEQRLFRERARAKEEELRESRARRAQEALGDREPKPTRAGPREEHPRGAGRGSLWRFGDVPWPCPGGGDPEAMAAALVARGPPLEEQGALRRYLRVQQVRWHPDRFLQRFRSQIETWELGRVMGAVTALSQALNRHAEALK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title