missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48847-25ul
This item is not returnable.
View return policy
Description
NFKBIL2 Polyclonal antibody specifically detects NFKBIL2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NFKBIL2 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| IkappaBR, I-kappa-B-related protein, IKBRFLJ40087, Inhibitor of kappa B-related protein, NF-kappa-B inhibitor-like protein 2, NFKBIL2tonsoku-like protein, Nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 2ikappaBR, tonsoku-like, DNA repair protein | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IRVRVQVQDHLFLIPVPHSSDTHSVAWLAEQAAQRYYQTCGLLPRLTLRKEGALLAPQDLIPDVLQSNDEVLAEVTSWDLPPLTDRYRRACQSLGQGEHQQVLQAVELQGLGLSFSACSLALDQAQLTPLLRALKLHT | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4796 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto