missing translation for 'onlineSavingsMsg'
Learn More

NFKBIZ Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. p-7223756 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16852474 20 μL 20µL
16842474 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16852474 Supplier Proteintech Supplier No. 140141AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The NFKBIZ antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NFKBIZ. This antibody recognizes human, mouse, rat antigen. The NFKBIZ antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Antigen NFKBIZ
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 307 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 64332
Target Species Human, Rat, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.