missing translation for 'onlineSavingsMsg'
Learn More

NGD 94-1, TRC

Product Code. 30477760
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477760 25 mg -
30466407 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477760 Supplier Toronto Research Chemicals Supplier No. TRCN39350825MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179333-18-7
Molecular Formula C18 H20 N6 . 2[C4 H4 O4]
Molecular Weight (g/mol) 552.536
Synonym NGD 94-1, CPDD 0045, NGD 941
IUPAC Name (Z)-but-2-enedioic acid;2-[4-[(2-phenyl-1H-imidazol-5-yl)methyl]piperazin-1-yl]pyrimidine
SMILES OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O.C(N1CCN(CC1)c2ncccn2)c3cnc([nH]3)c4ccccc4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H20N6.2C4H4O4/c1-2-5-15(6-3-1)17-21-13-16(22-17)14-23-9-11-24(12-10-23)18-19-7-4-8-20-18;2*5-3(6)1-2-4(7)8/h1-8,13H,9-12,14H2,(H,21,22);2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Formula Weight 552.197
Chemical Name or Material 2-[4-[(2-Phenyl-1H-imidazol-4-yl)methyl]-1-piperazinyl]-pyrimidine (2Z)-2-Butenedioate (1:2)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.