missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFI-B alpha/Nur77/NR4A1 Antibody (S02-1C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19726-100ul
This item is not returnable.
View return policy
Description
NGFI-B alpha/Nur77/NR4A1 Monoclonal antibody specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NGFI-B alpha/Nur77/NR4A1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S02-1C7 | |
| Unconjugated | |
| Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor | |
| A synthetic peptide of human NGFI-B alpha/Nur77/NR4A1 (Uniprot # P22736) | |
| 100 μg | |
| Apoptosis | |
| 3164 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction