missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR/TNFRSF16/p75NTR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00 € - 572.00 €
Specifications
| Antigen | NGFR/TNFRSF16/p75NTR |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407540
|
Novus Biologicals
NBP1-85769-25ul |
25 μL |
302.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18421331
|
Novus Biologicals
NBP1-85769 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NGFR/TNFRSF16/p75NTR Polyclonal antibody specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NGFR/TNFRSF16/p75NTR | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol | |
| 4804 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NKQGANSRPVNQTPPPEGEKLHSDSGISVDSQSLHDQQPHTQTASGQALKGDGGLYSSLPPAKREEVEKLLNGSAGDTWRHLAGELGYQPEHIDSFTHEACPVRALLASWATQDSATLDALLAALRRIQRADLVESLCSES | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title