missing translation for 'onlineSavingsMsg'
Learn More

NGI-1, MedChemExpress

Product Code. 16474878
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16474878

Brand: Medchem Express HY117383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NGI-1 (ML414) is a potent oligosaccharyltransferase (OST) inhibitor, directly targeting and blocking the function of the OST catalytic subunits STT3A and STT3B. NGI-1 is a cell permeable inhibitor and can effectively reduce virus infectivity without affecting cell viability.

NGI-1 (ML414) is a potent oligosaccharyltransferase (OST) inhibitor, directly targeting and blocking the function of the OST catalytic subunits STT3A and STT3B. NGI-1 is a cell permeable inhibitor and can effectively reduce virus infectivity without affecting cell viability.

Specifications

Chemical Name or Material NGI-1
CAS 790702-57-7
Color White
Purity Grade Notes Research
Molecular Formula C17H22N4O3S2
Quantity 10 mg
Synonym ML414
Solubility Information DMSO : 100 mg/mL (253.48 mM; Need ultrasonic)
SMILES O=C(NC1=NC=C(C)S1)C2=CC(S(=O)(N(C)C)=O)=CC=C2N3CCCC3
Molecular Weight (g/mol) 394.51
Formula Weight 394.51
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.