missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGLY1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94289-0.02ml
This item is not returnable.
View return policy
Description
NGLY1 Polyclonal antibody specifically detects NGLY1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| NGLY1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| EC 3.5.1.52, hPNGase, N-glycanase 1FLJ12409, Peptide:N-glycanase, peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase, PNG1FLJ11005, PNGase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human NGLY1 (NP_060767.2). MAAAALGSSSGSASPAVAELCQNTPETFLEASKLLLTYADNILRNPNDEKYRSIRIGNTAFSTRLLPVRGAVECLFEMGFEEGETHLIFPKKASVEQLQKIRDLIAIERSSRLDGSNKSHKVKSSQQPAASTQLPTTPSSNPSGLNQHTRNRQGQSSDPPSASTVAADSAILEVLQSNIQHVLVYENPALQEKALACIPVQELKRKSQEKLSRARKLDKGINISDEDFLLLELLHWFKEEFFHWVNNVLCSKCGGQTRSRDRSLLPSDDELKWGAKEVEDHYCDACQFSNRFPRYNNPEK | |
| 0.02 mL | |
| Cell Biology | |
| 55768 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction