missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGRN Mouse, anti-Human, Clone: 2C2C9, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | NGRN |
|---|---|
| Clone | 2C2C9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18095295
|
Proteintech
68593-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18070346
|
Proteintech
68593-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NGRN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NGRN. This antibody recognizes human antigen. The NGRN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NGRN | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| DSC92, FI58G, Neugrin, NGRN | |
| Primary |
| 2C2C9 | |
| Western Blot, ELISA | |
| Mouse | |
| 51335 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title