missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE3/SLC9A3, p Ser552 Antibody (14D5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 15 publications
227.00 € - 498.00 €
Specifications
| Antigen | NHE3/SLC9A3 (p Ser552) |
|---|---|
| Clone | 14D5 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1-5 ug/ml, Simple Western 1:1000, Flow Cytometry reported in scientific literature (PMID 21255452), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31276916), KnockDown Validated reported in scientific literature (PMID 31276916) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, KnockDown |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18780514
|
Novus Biologicals
NB110-81529SS |
0.025 mL |
227.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18791494
|
Novus Biologicals
NB110-81529 |
0.1 mL |
498.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated, Knockdown Validated.Specifications
| NHE3/SLC9A3 (p Ser552) | |
| 2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, KnockDown | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
| SLC9A3 | |
| IgG2a | |
| Protein G purified | |
| This antibody detects NHE3 posphorylated at Serine 552, and does not cross-react with the non-phosphopeptide. |
| 14D5 | |
| Western Blot 1-5 ug/ml, Simple Western 1:1000, Flow Cytometry reported in scientific literature (PMID 21255452), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31276916), KnockDown Validated reported in scientific literature (PMID 31276916) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
| 6550 | |
| Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 85 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title