missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93407-0.02ml
This item is not returnable.
View return policy
Description
NHE8 Polyclonal antibody specifically detects NHE8 in Human samples. It is validated for Western Blot
Specifications
| NHE8 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 472-581 of human SLC9A8 (NP_056081.1). RLMDIEDAKAHRRNKKDVNLSKTEKMGNTVESEHLSELTEEEYEAHYIRRQDLKGFVWLDAKYLNPFFTRRLTQEDLHHGRIQMKTLTNKWYEEVRQGPSGSEDDEQELL | |
| 0.02 mL | |
| Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism | |
| 23315 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction