missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHERF-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 572.00 €
Specifications
| Antigen | NHERF-2 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18417530
|
Novus Biologicals
NBP1-84944-25ul |
25ul |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18277997
|
Novus Biologicals
NBP1-84944 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NHERF-2 Polyclonal specifically detects NHERF-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NHERF-2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| E3KARP, Na(+)/H(+) exchange regulatory cofactor NHE-RF2, NHE3RF2, NHERF2MGC104639, NHERF-2NHE3 kinase A regulatory protein E3KARP, OCTS2, SIP-1SRY-interacting protein 1, sodium/hydrogen exchanger, Sodium-hydrogen exchanger regulatory factor 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 2, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2, Solute carrier family 9 isoform A3 regulatory factor 2, TKA-1SIP1, Tyrosine kinase activator protein 1 | |
| SLC9A3R2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q15599 | |
| 9351 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RLVEVNGVNVEGETHHQVVQRIKAVEGQTRLLVVDQETDEELRRRQLTCTEEMAQRGLPPAHDPWEPKPDWAHTGSHSSEAGKKDVSGPLRELRPRLCHLRKGPQGYGFNLH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 37 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title