missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicastrin Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
392.00 € - 578.00 €
Specifications
| Antigen | Nicastrin |
|---|---|
| Clone | 2 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261476
|
Novus Biologicals
NBP2-89825-50UL |
50 μg |
392.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18287176
|
Novus Biologicals
NBP2-89825-100UL |
100 μg |
578.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Flow Cytometry.Specifications
| Nicastrin | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
| Recombinant Human Nicastrin protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 23385 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title