missing translation for 'onlineSavingsMsg'
Learn More

Nickel - Ni @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 18011350
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18011350 50 g 50g
18082450 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18011350 Supplier VHG Labs Supplier No. VHGONI1000A50G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Nickel - Ni @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel
CAS 7440-02-0
Methods ICP/XRF/RDE
Analyte or Component Names Nickel
Shipping Condition Room Temperature
Molecular Formula Ni
Concentration 1000 μg/g
Certifications/Compliance ISO 17034
Synonym Raney alloy, Fibrex, Nickel catalyst, Nickel particles, Raney-Nickel
InChI Formula InChI=1S/Ni
SMILES [Ni]
IUPAC Name nickel
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 58.69
Quantity 50 g
Formula Weight 57.9353 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.