missing translation for 'onlineSavingsMsg'
Learn More

Nicotine N-Beta-D-Glucuronide, TRC

Product Code. 30007872
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007872 5 mg 5mg
30005158 10 mg 10mg
30009404 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007872 Supplier Toronto Research Chemicals Supplier No. TRCN4245755MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153536-53-9
Molecular Formula C16 H22 N2 O6
Molecular Weight (g/mol) 338.36
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S)-1-methyl-2-pyrrolidinyl]-, inner salt, Pyridinium, 1-β-D-glucopyranuronosyl-3-(1-methyl-2-pyrrolidinyl)-, inner salt, (S)-, Nicotine N-glucuronide, Nicotine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[3-[(2S)-1-methylpyrrolidin-2-yl]pyridin-1-ium-1-yl]oxane-2-carboxylate
SMILES CN1CCC[C@H]1c2ccc[n+](c2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H22N2O6/c1-17-6-3-5-10(17)9-4-2-7-18(8-9)15-13(21)11(19)12(20)14(24-15)16(22)23/h2,4,7-8,10-15,19-21H,3,5-6H2,1H3/t10-,11-,12-,13+,14-,15+/m0/s1
Formula Weight 338.1478
Percent Purity >95%
Chemical Name or Material Nicotine-N-beta-glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.