missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor gamma Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00 € - 470.00 €
Specifications
| Antigen | Nicotinic Acetylcholine Receptor gamma |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633131
|
Novus Biologicals
NBP2-93606-0.02ml |
0.02 mL |
204.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645011
|
Novus Biologicals
NBP2-93606-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine Receptor gamma Polyclonal antibody specifically detects Nicotinic Acetylcholine Receptor gamma in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Nicotinic Acetylcholine Receptor gamma | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, GPCR, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 1146 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| acetylcholine receptor subunit gamma, acetylcholine receptor, muscle, gamma subunit, ACHRG, cholinergic receptor, nicotinic, gamma, nicotinic, gamma polypeptide | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 328-517 of human CHRNG (NP_005190.4). NVSLRSPHTHSMARGVRKVFLRLLPQLLRMHVRPLAPAAVQDTQSRLQNGSSGWSITTGEEVALCLPRSELLFQQWQRQGLVAAALEKLEKGPELGLSQFCGSLKQAAPAIQACVEACNLIACARHQQSHFDNGNEEWFLVGRVLDRVCFLAMLSLFICGTAGIFLMAHYNRVPALPFPGDPRPYLPSPD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title