Learn More
Invitrogen™ Nidogen Monoclonal Antibody (ELM1)

Description
MA1-06501 detects entactin in human samples. MA1-06501 has sucessfully been used in Western blotting, immunosytochemistry and immunohistochemistry. By Western blotting, it detects a ~150 kDa protein representing entactin. The MA1-06501 immunogen is partially purified preparation of laminin from the EHS mouse tumor. Store at 4°C or in small aliquots at -20°C.
Specifications
Specifications
| Antigen | Nidogen |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | ELM1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | NID1 |
| Gene Accession No. | P10493 |
| Gene Alias | A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.