missing translation for 'onlineSavingsMsg'
Learn More

Nifekalant Hydrochloride, TRC

Product Code. 17942885
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942885 1 g 1g
17952025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942885 Supplier Toronto Research Chemicals Supplier No. TRCN4571001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130656-51-8
Molecular Formula C19 H27 N5 O5 . Cl H
Molecular Weight (g/mol) 441.9091
Synonym 6-[[2-[(2-Hydroxyethyl)[3-(4-nitrophenyl)propyl]amino]ethyl]amino]-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione Hydrochloride, MS 551, Shinbit
IUPAC Name 6-[2-[2-hydroxyethyl-[3-(4-nitrophenyl)propyl]amino]ethylamino]-1,3-dimethylpyrimidine-2,4-dione;hydrochloride
SMILES Cl.CN1C(=O)C=C(NCCN(CCO)CCCc2ccc(cc2)[N+](=O)[O-])N(C)C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H27N5O5.ClH/c1-21-17(14-18(26)22(2)19(21)27)20-9-11-23(12-13-25)10-3-4-15-5-7-16(8-6-15)24(28)29;/h5-8,14,20,25H,3-4,9-13H2,1-2H3;1H
Formula Weight 441.1779
Chemical Name or Material Nifekalant Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.