missing translation for 'onlineSavingsMsg'
Learn More

Nifursol (~90%), TRC

Product Code. 30137888
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137888 50 mg 50mg
30132750 5 mg 5mg
17922355 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137888 Supplier Toronto Research Chemicals Supplier No. TRCN45870050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16915-70-1
Molecular Formula C12 H7 N5 O9
Molecular Weight (g/mol) 365.21
Synonym 2-Hydroxy-3,5-dinitrobenzoic acid 2-[(5-nitro-2-furanyl)methylene]hydrazide, 2-Hydroxy-3,5-dinitrobenzoic acid [(5-nitro-2-furanyl)methylene]hydrazide, 3,5-Dinitrosalicylic acid (5-nitrofurfurylidene)hydrazide, Histomon, Nifursol, Salfuride, Sulfuride
IUPAC Name 2-hydroxy-3,5-dinitro-N-[(E)-(5-nitrofuran-2-yl)methylideneamino]benzamide
SMILES Oc1c(cc(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N\N=C\c2oc(cc2)[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H7N5O9/c18-11-8(3-6(15(20)21)4-9(11)16(22)23)12(19)14-13-5-7-1-2-10(26-7)17(24)25/h1-5,18H,(H,14,19)/b13-5+
Formula Weight 365.0244
Chemical Name or Material Nifursol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.