missing translation for 'onlineSavingsMsg'
Learn More

Nigericin sodium salt, MedChemExpress

Product Code. 16485717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485717 5 mg 5mg
16469546 50 mg 50mg
16462978 10 mg 10mg
16452566 25 mg 25mg
16444296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485717 Supplier Medchem Express Supplier No. HY100381
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator.

Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator.

Specifications

Chemical Name or Material Nigericin sodium salt
Physical Form Powder
CAS 28643-80-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C40H67NaO11
For Use With (Application) Cancer-programmed cell death
Solubility Information Ethanol : ≥ 50 mg/mL (66.94 mM)
SMILES C[C@H]1[C@](O[C@@H]2C[C@](CC[C@@H]3C)([H])O[C@@]3([H])[C@@H](C)C(O[Na])=O)([C@@H]([C@H](OC)C2)C)O[C@](C)([C@@]4([H])O[C@@](C)([C@@]5([H])O[C@@]([C@]([C@@H](C[C@@H]6C)C)([H])O[C@]6(O)CO)([H])C[C@H]5C)CC4)C1
Molecular Weight (g/mol) 746.94
Formula Weight 746.94
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.