missing translation for 'onlineSavingsMsg'
Learn More

Nigericin sodium salt, MedChemExpress

Product Code. 16444296
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444296

Brand: Medchem Express HY100381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator.

Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator.

Specifications

Chemical Name or Material Nigericin sodium salt
Physical Form Powder
CAS 28643-80-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C40H67NaO11
For Use With (Application) Cancer-programmed cell death
Solubility Information Ethanol : ≥ 50 mg/mL (66.94 mM)
SMILES C[C@H]1[C@](O[C@@H]2C[C@](CC[C@@H]3C)([H])O[C@@]3([H])[C@@H](C)C(O[Na])=O)([C@@H]([C@H](OC)C2)C)O[C@](C)([C@@]4([H])O[C@@](C)([C@@]5([H])O[C@@]([C@]([C@@H](C[C@@H]6C)C)([H])O[C@]6(O)CO)([H])C[C@H]5C)CC4)C1
Molecular Weight (g/mol) 746.94
Formula Weight 746.94
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.