missing translation for 'onlineSavingsMsg'
Learn More

Nilotinib Regio Isomer, TRC

Product Code. 30006474
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006474 50 mg 50mg
30005482 5 mg 5mg
30004023 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006474 Supplier Toronto Research Chemicals Supplier No. TRCN46532050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 641571-15-5
Molecular Formula C28 H22 F3 N7 O
Molecular Weight (g/mol) 529.52
Synonym 4-methyl-N-[3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, 4-Methyl-N-[3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]benzamide, 4-Methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-N-[5-(5-methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)phenyl]benzamide, 4-Methyl-N-[3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzamide, Nilotinib isomer
IUPAC Name 4-methyl-N-[3-(5-methylimidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]benzamide
SMILES Cc1ccc(cc1Nc2nccc(n2)c3cccnc3)C(=O)Nc4cc(cc(c4)C(F)(F)F)n5cncc5C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H22F3N7O/c1-17-5-6-19(10-25(17)37-27-34-9-7-24(36-27)20-4-3-8-32-15-20)26(39)35-22-11-21(28(29,30)31)12-23(13-22)38-16-33-14-18(38)2/h3-16H,1-2H3,(H,35,39)(H,34,36,37)
Formula Weight 529.1838
Chemical Name or Material Nilotinib regioisomer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.