missing translation for 'onlineSavingsMsg'
Learn More

Nimustine hydrochloride, MedChemExpress

Product Code. 16491967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491967 50 mg 50mg
16476286 100 mg 100mg
16445818 10 mg 10mg
16408487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491967 Supplier Medchem Express Supplier No. HY13703A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nimustine hydrochloride (ACNU) is a DNA cross-linking and DNA alkylating agent, which induces DNA replication blocking lesions and DNA double-strand breaks and inhibits DNA synthesis, commonly used in chemotherapy for glioblastomas.

Nimustine hydrochloride (ACNU) is a DNA cross-linking and DNA alkylating agent, which induces DNA replication blocking lesions and DNA double-strand breaks and inhibits DNA synthesis, commonly used in chemotherapy for glioblastomas.

Specifications

Chemical Name or Material Nimustine hydrochloride
Physical Form Solid
CAS 55661-38-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C9H14Cl2N6O2
For Use With (Application) Cancer-programmed cell death
Synonym ACNU
Solubility Information DMSO : 62.5 mg/mL (202.17 mM; Need ultrasonic)
SMILES O=C(NCC1=CN=C(C)N=C1N)N(CCCl)N=O.[H]Cl
Molecular Weight (g/mol) 309.15
Formula Weight 309.15
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.