missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ninjurin 1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15981475
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15981475 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15981475 Supplier Invitrogen™ Supplier No. PA572821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ninjurin 1 Polyclonal Antibody for ELISA, IHC (P)

Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues. Cell adhesion requires divalent cations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninjurin 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ninj1
Gene Accession No. O70131, P70617, Q92982
Gene Alias AU024536; Nerve injury-induced protein 1; nerve injury-induced protein-1; NIN1; Ninj1; NINJURIN; ninjurin 1; ninjurin-1
Gene Symbols Ninj1
Host Species Rabbit
Immunogen NINJ1 antibody for a 15 amino acid peptide near the carboxy terminus of human NINJ1. The immunogen is within the last 50 amino acids of NINJ1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18081, 25338, 4814
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.