missing translation for 'onlineSavingsMsg'
Learn More

Nipecotamide, 95%, Thermo Scientific Chemicals

Product Code. 10153180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153180 5 g Glass Bottle 5g
10466042 50 g Glass Bottle 50g
10732374 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153180 Supplier Thermo Scientific Acros Supplier No. 128300050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4138-26-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
MDL Number MFCD00005993
InChI Key BVOCPVIXARZNQN-UHFFFAOYSA-N
Synonym nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide
PubChem CID 92980
ChEBI CHEBI:60116
IUPAC Name piperidine-3-carboxamide
SMILES C1CC(CNC1)C(=O)N

Specifications

Melting Point 102°C to 108°C
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 128.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Nipecotamide, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.