missing translation for 'onlineSavingsMsg'
Learn More

NISCH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16862204 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16862204 150 μL 150µL
16872204 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16862204 Supplier Proteintech Supplier No. 138131AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Acts either as the functional imidazoline-1 receptor (I1R) candidate or as a membrane-associated mediator of the I1R signaling. Binds numerous imidazoline ligands that induces initiation of cell-signaling cascades triggering to cell survival, growth and migration. Its activation by the agonist rilmenidine induces an increase in phosphorylation of mitogen-activated protein kinases MAPK1 and MAPK3 in rostral ventrolateral medulla (RVLM) neurons that exhibited rilmenidine-evoked hypotension. Blocking its activation with efaroxan abolished rilmenidine-induced mitogen-activated protein kinase phosphorylation in RVLM neurons. Acts as a modulator of Rac-regulated signal transduction pathways. Suppresses Rac1-stimulated cell migration by interacting with PAK1 and inhibiting its kinase activity. Also blocks Pak-independent Rac signaling by interacting with RAC1 and inhibiting Rac1-stimulated NF-kB response element and cyclin D1 promoter activation. Inhibits also LIMK1 kinase activity by reducing LIMK1 ′Tyr-508′ phosphorylation. Inhibits Rac-induced cell migration and invasion in breast and colon epithelial cells. Inhibits lamellipodia formation, when overexpressed. Plays a role in protection against apoptosis. Involved in association with IRS4 in the enhancement of insulin activation of MAPK1 and MAPK3. When overexpressed, induces a redistribution of cell surface ITGA5 integrin to intracellular endosomal structures.

Specifications

Antigen NISCH
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NISCH
Gene Accession No. Q4G017, Q80TM9, Q9Y2I1
Gene Alias hIRAS, I 1, I 1 receptor candidate protein, I1R, I1R candidate protein, Imidazoline 1 receptor, Imidazoline receptor 1, IR1, IRAS, KIAA0975, NISCH, nischarin
Gene Symbols Nisch
Host Species Rabbit
Immunogen NISCH Fusion Protein Ag4880
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11188, 306255, 64652
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.