missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Concentrate, 100ppm, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11444769 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444769 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11444769 Supplier Reagecon Supplier No. 5002100C
  • ONLINE EXCLUSIVE PRICE
    187.25 €
    142.37 € / 100mL
    Save 44.88 €
    23% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Concentrated to make nitrate standard solution. Nitrate Concentrate, 100ppm, EP Grade, Reagecon™ can be used to make a solution with 100ppm NO3. It is manufactured according to European Pharmacopoeia Chapter 4(4.1.2) for use in nitrate determination and limit tests.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4(4.1.2) guidelines
  • Produced in accordance with ASTM, APHA, ACS, EP and USP methods
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use solution eliminates the time and labor of preparing precise standard solutions

Specifications

Packaging 100mL
CAS 14797-55-8
Certificate of Analysis Yes
Odor Odorless
Physical Form Liquid
Molecular Formula NO3-
Concentration 100 ppm NO3
Color Colorless
InChI Key NHNBFGGVMKEFGY-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-]
IUPAC Name nitrate
Molecular Weight (g/mol) 62.004
PubChem CID 943
Quantity 100 mL
ChEBI CHEBI:17632
Formula Weight 62.00g/mol
Grade Eu. Ph. Chapter 4(4.1.2)
Chemical Name or Material Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.