missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride, 90%

Product Code. p-4813583 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 10289450

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.65
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]

Specifications

Melting Point 189°C
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 817.65
Percent Purity 90%
Chemical Name or Material Nitro Blue Tetrazolium Chloride
compliance-icons
Product Identifier
  • Nitro Blue Tetrazolium Chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2H-Tetrazolium, 3,3-(3,3-dimethoxy[1,1-biphenyl]-4,4-diyl)bis[2-(4-nitrophenyl)-5-phenyl-, dichlori

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.